Hard Plastic Microscope Slide Tray, Electron Microscopy Sciences
À propos de cet article
This durable ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1”).
Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable. Measures 11 7/8” L x 7 ”W x ½”H (300 x 195 x 12 mm).
2 Options Available Below
Product Details & Documents
This durable ABS hard plastic slide tray will hold up to 20 standard microscope slides 75 x 25 mm (3x1”).
Ivory color consists of two rows, each with securely holds 10 slides. Each numbered cavity has thumb cut and raise notches for easy to remove the slide. Stackable. Measures 11 7/8” L x 7 ”W x ½”H (300 x 195 x 12 mm).
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 71503-01
- Hard Plastic Microscope Slide TrayEach$179.70Pack of 10$126.40Supplier Number: 71503-10