Slide Box-Foam, Electron Microscopy Sciences
À propos de cet article
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
- Holds 25, 50, or 100 slides
- Foam backing
- Secure lock
- Blue, green, red, yellow, white, black, and pink
11 Options Available Below
Product Details & Documents
Slide Box with a foam backing and features a secure lock to ensure it does not open accidently.
- Holds 25, 50, or 100 slides
- Foam backing
- Secure lock
- Blue, green, red, yellow, white, black, and pink
Product Family Options
Informations sur le produit
- DésignationCapacitéColorAvailabilityPrice
- Supplier Number: 71478-01Spécifications:
More
Specifications
Cat. No.102097-542 - Supplier Number: 71478-02Spécifications:
More
Specifications
Cat. No.102097-544 - Slide Box25RedDiscontinued-Supplier Number: 71478-03Spécifications:
More
Specifications
Cat. No.102097-546 - Slide Box25YellowDiscontinued-Supplier Number: 71478-04Spécifications:
More
Specifications
Cat. No.102097-548 - Slide Box25WhiteDiscontinued-Supplier Number: 71478-05Spécifications:
More
Specifications
Cat. No.102097-550 - Supplier Number: 71478-06Spécifications:
More
Specifications
Cat. No.102097-552 - Slide Box50GreenDiscontinued-Supplier Number: 71478-08Spécifications:
More
Specifications
Cat. No.102097-556 - Slide Box50RedDiscontinued-Supplier Number: 71478-09Spécifications:
More
Specifications
Cat. No.102097-558 - Slide Box50YellowDiscontinued-Supplier Number: 71478-10Spécifications:
More
Specifications
Cat. No.102097-560 - Supplier Number: 71478-11Spécifications:
More
Specifications
Cat. No.102097-562 - Supplier Number: 71478-12Spécifications:
More
Specifications
Cat. No.102097-564 - Supplier Number: 71478-13Spécifications:
More
Specifications
Cat. No.102097-566 - Supplier Number: 71478-14Spécifications:
More
Specifications
Cat. No.102097-568 - Supplier Number: 71478-15Spécifications:
More
Specifications
Cat. No.102097-570 - Supplier Number: 71478-16Spécifications:
More
Specifications
Cat. No.102097-572 - Supplier Number: 71478-17Spécifications:
More
Specifications
Cat. No.102097-574 - Supplier Number: 71478-18Spécifications:
More
Specifications
Cat. No.102097-576