Grid Transporting Box, Electron Microscopy Sciences | Avantor
Grid Transporting Box, Electron Microscopy Sciences
Grid Transporting Box
Catalog # 102097-450
Fournisseur #:  71173-01
Grid Transporting Box, Electron Microscopy Sciences
Catalog # 102097-450
Grid Transporting Box, Electron Microscopy Sciences
Grid Transporting Box, Electron Microscopy Sciences
Grid Transporting Box
Price
$201.69
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This unique Box allows for the storage and transportation of grids without any worry of them moving around or being damaged.
The Plate is made from Silicone and has 54 individual compartments 10 mm in size. Each compartment can hold 3 grids and the grids can be easily picked up from the Silicone surface.
Specifications
  • Désignation:
    Grid Transporting Box