Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
Micro Tissue Grinder Kit, Electron Microscopy Sciences
This complete selection of Wheaton Micro Tissue Grinders is conveniently packed in a lightweight protective carrying case. The 0.5 ml micro tissue grinder with screw cap can be used for tissue grinding as well as for additional procedures.
  • Complete kit includes 7 micro tissue grinders
  • Made from Wheaton 33 low extractable borosilicate glass
  • Replacement parts available separately
  • Autoclavable
This particular tissue grinder mortar can be safely joined with another threaded glass component if needed, using the Wheaton Connection®. The Kit is packed in high-density polyethylene case with foam inserts.
Product Family Options
  • Désignation
    Taille
  • Complete Kit
    Each
    Each - 2 019,97 $
    2 019,97 $
    Each
       2 019,97 $
     : 64800
  • Tissue Grinder
    0.2 ml
    Each
    Each - 423,99 $
    423,99 $
    Pack of 2
    Pack of 2 - 298,86 $
    298,86 $
    Each
       423,99 $
    Pack of 2
       298,86 $
     : 64799-02