Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Certificates
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All features are similar to the Potter-Elvehjem Tissue Grinder series, with additional radial serrated PTFE pestle.
This grinder has a serrated PTFE pestle that disperses homogenate into the mortar cylinder more efficiently. Depth of the serration is 0.1 to 0.15 mm (0.20 to 0.25”).
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Tissue Grinder PTFE with Radial Serrations
    2 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $778.33
    $778.33
    Log in pour la disponibilité
    Pack of 2
    Pack of 2 - $547.53
    $547.53
    Each
       $778.33
    Pack of 2
       $547.53
    Supplier Number: 64795-02
  • Tissue Grinder PTFE with Radial Serrations
    5 ml
    Discontinued
    Supplier Number: 64795-05
  • Tissue Grinder PTFE with Radial Serrations
    10 ml
    Discontinued
    Supplier Number: 64795-10
  • Tissue Grinder With Radial Serrations
    15 ml
    Discontinued
    Supplier Number: 64795-15
  • Tissue Grinder PTFE with Radial Serrations
    30 ml
    Discontinued
    Supplier Number: 64795-30
  • Tissue Grinder PTFE with Radial Serrations
    55 ml
    Discontinued
    Supplier Number: 64795-55
  • Tissue Grinder Pestle only PTFE Radial Serrations
    2 ml
    Discontinued
    Supplier Number: 64795-80
  • Tissue Grinder Pestle only PTFE Radial Serrations
    5 ml
    Discontinued
    Supplier Number: 64795-81
  • Tissue Grinder Pestle only PTFE Radial Serrations
    10 ml
    Discontinued
    Supplier Number: 64795-82
  • Tissue Grinder Pestle only PTFE Radial Serrations
    15 ml
    Discontinued
    Supplier Number: 64795-83
  • Tissue Grinder Pestle only PTFE Radial Serrations
    30 ml
    Discontinued
    Supplier Number: 64795-84
  • Tissue Grinder Pestle only PTFE Radial Serrations
    55 ml
    Discontinued
    Supplier Number: 64795-85