Potter-Elvehjem Tissue Grinder, Electron Microscopy Sciences
About this item
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. They are designed for use in preparation of tissue homogenates and other similar applications.
- Ideal for tissue homogenates
- Use with power homogenization
- Parts interchangeable
- Teflon® pestle
- Autoclavable
The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3mm) diameter. The grinding chamber clearance is 0.004" to 0.006’ (0.1 to 0.15mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer. However, we strongly recommend the use of safety-coated tissue grinder (64793-series) for added safety. This unit is manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type 1 and ASTM E-438 Type 1, Class A requirements.
1 Options Available Below
Product Details & Documents
These grinder are precisely constructed with completely interchangeable parts to allow easier cleaning or replacement of damaged mortars. They are designed for use in preparation of tissue homogenates and other similar applications.
- Ideal for tissue homogenates
- Use with power homogenization
- Parts interchangeable
- Teflon® pestle
- Autoclavable
The Teflon pestle is for soft tissues such as brain or liver. The stainless steel rod is 0.25" (6.3mm) diameter. The grinding chamber clearance is 0.004" to 0.006’ (0.1 to 0.15mm). This unit is autoclavable and can be used for power homogenization as with overhead stirrer. However, we strongly recommend the use of safety-coated tissue grinder (64793-series) for added safety. This unit is manufactured from Wheaton 33 low extractable borosilicate glass that conforms to USP Type 1 and ASTM E-438 Type 1, Class A requirements.
Product Family Options
Product Information
- DésignationTailleAvailabilityPrice
- Tissue Grinder2 mlDiscontinued-Supplier Number: 64792-02
- Tissue Grinder5 mlDiscontinued-Supplier Number: 64792-05
- Tissue Grinder10 mlDiscontinued-Supplier Number: 64792-10
- Tissue Grinder15 mlDiscontinued-Supplier Number: 64792-15
- Tissue Grinder30 mlDiscontinued-Supplier Number: 64792-30
- Tissue Grinder55 mlEach$1,181.37Pack of 2$868.91Supplier Number: 64792-55
- Mortar only Tissue Grinder2 mlDiscontinued-Supplier Number: 64792-70
- Mortar only Tissue Grinder5 mlDiscontinued-Supplier Number: 64792-71
- Mortar only Tissue Grinder10 mlDiscontinued-Supplier Number: 64792-72
- Mortar only Tissue Grinder15 mlDiscontinued-Supplier Number: 64792-73
- Mortar only Tissue Grinder30 mlDiscontinued-Supplier Number: 64792-74
- Mortar only Tissue Grinder55 mlDiscontinued-Supplier Number: 64792-75
- Pestle only Tissue Grinders2 mlDiscontinued-Supplier Number: 64792-80
- Pestle only Tissue Grinders5 mlDiscontinued-Supplier Number: 64792-81
- Pestle only Tissue Grinders10 mlDiscontinued-Supplier Number: 64792-82
- Pestle only Tissue Grinders15 mlDiscontinued-Supplier Number: 64792-83
- Pestle only Tissue Grinders30 mlDiscontinued-Supplier Number: 64792-84
- Pestle only Tissue Grinders55 mlDiscontinued-Supplier Number: 64792-85