Frozen Section Brush, Electron Microscopy Sciences | Avantor
Frozen Section Brush, Electron Microscopy Sciences
Frozen Section Brushes, set of 4
Catalog # 102093-794
Fournisseur #:  61735-01
Frozen Section Brush, Electron Microscopy Sciences
Catalog # 102093-794
Frozen Section Brush, Electron Microscopy Sciences
Frozen Section Brush, Electron Microscopy Sciences
Frozen Section Brushes, set of 4
Price
$150.58
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The stiffness of this bristle will prevent the frozen section from curling through the brush. The flat white boar bristle brush is cut into an angle.
Set of four brushes: two #1 with ⅜" angle cut and #2 with ¼" angle cut.
Spécifications
  • Désignation:
    Frozen Section Brushes, set of 4