Foil Roll, Electron Microscopy Sciences
À propos de cet article
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
- Create custom-size pieces of foil
- Minimal waste
- Two sizes available
Designed to work without Mini Bin™ Foil Dispenser, these foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing and general storage.
2 Options Available Below
Product Details & Documents
Custom-sized pieces of foil can easily be created for covering small surfaces such as beakers, flasks, tubes, and petri dishes.
- Create custom-size pieces of foil
- Minimal waste
- Two sizes available
Designed to work without Mini Bin™ Foil Dispenser, these foil rolls are ideal for autoclaving, freezing, incubating, protecting lab ware, lining, sealing and general storage.