DURAX™ Tweezers, Electron Microscopy Sciences
À propos de cet article
With a 616HV Vickers-hardness, the tip of Rubis DURAX™ tweezers is six times harder than the tip of a standard steel tweezers.
- Higher resistance against normal wear and tear
- Longer service life
Even with intensive use, these tweezers retain their original precision. Their specially-hardened chrome-magnesium steel is also used in the manufacture of surgicals.
4 Options Available Below
Product Details & Documents
With a 616HV Vickers-hardness, the tip of Rubis DURAX™ tweezers is six times harder than the tip of a standard steel tweezers.
- Higher resistance against normal wear and tear
- Longer service life
Even with intensive use, these tweezers retain their original precision. Their specially-hardened chrome-magnesium steel is also used in the manufacture of surgicals.
Product Family Options
Informations sur le produit
- DésignationOverall LengthTip LengthTip OpeningTip WidthAvailabilityPrice
- Style 3C110 mm34 mm1.5 mm9 mmDiscontinued-Supplier Number: 78149-3C
- Style 4110 mm33 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-4
- Supplier Number: 78149-0C
- Supplier Number: 78149-5
- Style 5115 mm30 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-5A
- Supplier Number: 78149-7E
- Style 0120 mm82 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-0
- Style 00120 mm50 mm1.5 mm9 mmDiscontinued-Supplier Number: 78149-00
- Style 0C 11120 mm50 mm1.5 mm9 mmDiscontinued-Supplier Number: 78149-0C11
- Style 1120 mm43 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-1
- Style 2120 mm35 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-2
- Supplier Number: 78149-2A
- Style 7120 mm34 mm1.5 mm11 mmDiscontinued-Supplier Number: 78149-7
- Style F120 mm33 mm1.5 mm10 mmDiscontinued-Supplier Number: 78149-F
- Style SS135 mm47 mm1.5 mm8 mmDiscontinued-Supplier Number: 78149-SS