Swiss Tweezer; Precision, Electron Microscopy Sciences
À propos de cet article
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
- Tweezers engineered for a NON-SLIP GRIP
- Light weight –prevents hand fatigue
- Sensuous touch and functional design
- Ultra-fine precision tips
- Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.
6 Options Available Below
Product Details & Documents
Swiss precision tweezer made from austenitic, acid-proof stainless steel, 100% anti-magnetic (SA).
- Tweezers engineered for a NON-SLIP GRIP
- Light weight –prevents hand fatigue
- Sensuous touch and functional design
- Ultra-fine precision tips
- Melting point 1400°C
Resistant to hydrogen fluoride (HF) and nitric acid.
Product Family Options
Informations sur le produit
- DésignationLongueurTip DimensionsAvailabilityPrice
- Supplier Number: 78170-1G
- Supplier Number: 78170-2AG
- Supplier Number: 78170-2G
- Supplier Number: 78170-3CG
- Style 3G120 mmM=10 mm; N = 1.5 mm; P= 50 mmDiscontinued-Supplier Number: 78170-3G
- Style 4G110 mmM= 10 mm; N = 1.5 mmDiscontinued-Supplier Number: 78170-4G
- Style 5AGDiscontinued-Supplier Number: 78170-5AG
- Style 5ARG115 mmM= 10 mm; N = 1.5 mmDiscontinued-Supplier Number: 78170-5ARG
- Supplier Number: 78170-5G
- Supplier Number: 78170-7G