Micro Scissors, Rubis, Electron Microscopy Sciences
About this item
Micro scissors manufactured from hardened, anti-acid, stainless, surgical steel (CA)
Rubis Micro Scissors Set includes one light duty Rubis Colibri fine scissors, one medium duty Rubis Classic medium fine scissors, and one heavy duty Rubis Sauro medium strong scissors. Strong, slightly curved easily cut through small bones and cartilages.
Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist® grip scissors offer ergonomically, comfort and control.
2 Options Available Below
Product Details & Documents
Micro scissors manufactured from hardened, anti-acid, stainless, surgical steel (CA)
Rubis Micro Scissors Set includes one light duty Rubis Colibri fine scissors, one medium duty Rubis Classic medium fine scissors, and one heavy duty Rubis Sauro medium strong scissors. Strong, slightly curved easily cut through small bones and cartilages.
Handcraft with precision sharpened blades that provide a perfect cut. The Rubis Twist® grip scissors offer ergonomically, comfort and control.
Product Family Options
Product Information
- DésignationLongueurBlade LengthAvailabilityPrice
- Style Micro Scissors, 1C300110 mm (4⅓")5.4 mm (¼")Discontinued-Supplier Number: 78180-1C3
- Supplier Number: 78180-1C2
- Supplier Number: 78180-03