Dual Micrometer Calibration Slide, Electron Microscopy Sciences
À propos de cet article
EMS has introduced two new calibration slides that have the benefit of dual imperial/metric scales.
- New Dual-Scale Calibration Slides
- 2" Imperial (English) and 50mm Metric Scales on a Single Slide
- Positive and Negative Versions
- Unique Serial Number for Traceability
- Available with Internationally Traceable Certificates of Calibration
The PS52P is for transmitted light applications and has a bright chrome positive image. The PS52N has a negative pattern, formed in low reflective chrome for incident light applications to give excellent contrast. Both are ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Standard and Crossed Patterns – S1, S2, S4, S5, S8, S11, S12, S16, S20, S21, S22, S48.
2 Options Available Below
Product Details & Documents
EMS has introduced two new calibration slides that have the benefit of dual imperial/metric scales.
- New Dual-Scale Calibration Slides
- 2" Imperial (English) and 50mm Metric Scales on a Single Slide
- Positive and Negative Versions
- Unique Serial Number for Traceability
- Available with Internationally Traceable Certificates of Calibration
The PS52P is for transmitted light applications and has a bright chrome positive image. The PS52N has a negative pattern, formed in low reflective chrome for incident light applications to give excellent contrast. Both are ideal for calibrating optical products with a large field of view, such as stereo microscopes or imaging systems. Standard and Crossed Patterns – S1, S2, S4, S5, S8, S11, S12, S16, S20, S21, S22, S48.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Dual micrometer calibration slides PS52P with ukas certificateDiscontinued-Supplier Number: 68040-06
- Dual micrometer calibration slides PS52N with ukas certificateDiscontinued-Supplier Number: 68040-08
- Dual micrometer calibration slide, 76 mm×25 mm, positive imLog in pour la disponibilitéEach$1,598.12Supplier Number: 68040-05
- Dual micrometer calibration slide, 76 mm×25 mm, negative imaLog in pour la disponibilitéEach$1,954.72Supplier Number: 68040-07