Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences | Avantor
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Certificates
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
Harris Uni-Core™ Sampling Tool, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Harris Uni-Core™ is a disposable multi-purpose sampling tool for soft substrates. Made from polypropylene, including a hollow body with built-in tool steel razor sharp cutting tip, with a plunger for the expelling of the punched sample.
  • Individually wrapped
  • Sterilized with ethylene oxide
  • Reuse by rinsing the tip with ethanol and autoclaving for 20 minutes at 250 °F/15 psi
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Sampling Tool 1.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $103.73
    $103.73
    Each
       $103.73
    Supplier Number: 69036-10
  • Sampling Tool 3.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $75.60
    $75.60
    Each
       $75.60
    Supplier Number: 69036-30
  • Sampling Tool 6.0
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $71.08
    $71.08
    Each
       $71.08
    Supplier Number: 69036-60