Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
À propos de cet article
Head: 32 mm (11/4”) and Height: 6 mm (1/4”).
Head: 32 mm (1 1/4”). Height: 6 mm (1/4”).
1 Options Available Below
Product Details & Documents
Head: 32 mm (11/4”) and Height: 6 mm (1/4”).
Head: 32 mm (1 1/4”). Height: 6 mm (1/4”).
Product Family Options
Informations sur le produit
- DésignationMatièreAvailabilityPrice
- Hitachi M4AluminumEach$84.68Pack of 10$70.01Supplier Number: 75660
- Hitachi M4AluminumDiscontinued-Supplier Number: 75661
- Hitachi M4AluminumDiscontinued-Supplier Number: 75662