Hitachi M4 Thread SEM Mount, Electron Microscopy Sciences
About this item
4 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- DésignationMatièreHead DiameterHauteurAvailabilityPrice
- Hitachi M4Aluminum15 mm (⅝")10 mm (⅜")Discontinued-Supplier Number: 75635-10
- Hitachi M4Aluminum15 mm (⅝")10 mm (⅜")Discontinued-Supplier Number: 75635-20
- Hitachi M4Aluminum15 mm (⅝")10 mm (⅜")Discontinued-Supplier Number: 75635-30
- Hitachi M4Aluminum25 mm (1")10 mm (⅜")Discontinued-Supplier Number: 75635-40
- Hitachi M4Aluminum25 mm (1")10 mm (⅜")Discontinued-Supplier Number: 75635-50
- Hitachi M4Aluminum25 mm (1")10 mm (⅜")Discontinued-Supplier Number: 75635-60
- Hitachi M4 12 divisionsAluminum32 mm (1¼")6.0 mm (¼")Discontinued-Supplier Number: 75635-70D
- Hitachi M4Aluminum32 mm (1¼")6.0 mm (¼")Each119,56 $Pack of 1098,92 $Supplier Number: 75635-70
- Hitachi M4Aluminum32 mm (1¼")10 mm (⅜")Discontinued-Supplier Number: 75635-75
- Hitachi M4Aluminum50 mm (2")6.0 mm (¼")Each209,88 $Pack of 10175,05 $Supplier Number: 75635-77
- Hitachi M4 12 divisionsAluminum32 mm (1¼")6.0 mm (¼")Discontinued-Supplier Number: 75635-80D
- Hitachi M4Aluminum32 mm (1¼")6.0 mm (¼")Discontinued-Supplier Number: 75635-80
- Hitachi M4Aluminum32 mm (1¼")10 mm (⅜")Each401,84 $Pack of 50336,08 $Supplier Number: 75635-85
- Hitachi M4Aluminum50 mm (2")6.0 mm (¼")Discontinued-Supplier Number: 75635-87
- Hitachi M4 12 divisionsAluminum32 mm (1¼")6.0 mm (¼")Discontinued-Supplier Number: 75635-90D
- Hitachi M4Aluminum32 mm (1¼")6.0 mm (¼")Each724,16 $Pack of 100606,83 $Supplier Number: 75635-90
- Hitachi M4Aluminum32 mm (1¼")10 mm (⅜")Discontinued-Supplier Number: 75635-95
- Hitachi M4Aluminum50 mm (2")6.0 mm (¼")Discontinued-Supplier Number: 75635-97