Forensic Certified Mount, Electron Microscopy Sciences
À propos de cet article
Gun Shot residue mount is free from objectional elements that would interfere with results.
- Made from special certified aluminum alloy
- Mounts are 12.7 x 11 mm
- Available with 8 or 6 mm pin
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR.
3 Options Available Below
Product Details & Documents
Gun Shot residue mount is free from objectional elements that would interfere with results.
- Made from special certified aluminum alloy
- Mounts are 12.7 x 11 mm
- Available with 8 or 6 mm pin
This mount is made from a special certified Aluminum alloy which does not contain any of the objectionable elements that may interfere with samples collected for GSR.
Product Family Options
Informations sur le produit
- DésignationPin SizeAvailabilityPrice
- SEM Mount8 mmEach$12.31Pack of 10$11.22Supplier Number: 76475-05
- SEM Mount8 mmDiscontinued-Supplier Number: 76475-10
- SEM Mount8 mmDiscontinued-Supplier Number: 76475-50
- Supplier Number: 76475-52
- Supplier Number: 76475-54