Single and Multi Wafer Containers, Electron Microscopy Sciences
À propos de cet article
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
- Conical base to support the wafers on their edges
- Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
: The set includes the container, the cover and the spring.
9 Options Available Below
Product Details & Documents
These containers are for storage and shipping of a variety of sizes of wafers (Any material) from 1 to 6" in diameter. They prevent damage and contamination to the wafer. Features a conical base to support the wafers on their edges. Wafers are held in place by a spring.
- Conical base to support the wafers on their edges
- Wafers are held in place by the spring
The multi-wafer containers are made from polypropylene and have a storage capacity of 25 wafers maximum. They come complete with the base and cover. The single containers will accommodate a variety of sizes from 4 to 6" by simply adjusting the silicone supports in the box. These containers are made from anti-electrostatic polycarbonate and are available in transparent or black.
: The set includes the container, the cover and the spring.
Product Family Options
Informations sur le produit
- DésignationTailleFor Use WithAvailabilityPrice
- Wafer Container Single33 x 10 mm25 mm (1")Each$85.86Pack of 6$65.06Supplier Number: 64917-1
- Wafer Container Single60 x 11 mm50 mm (2")Each$133.77Pack of 6$102.10Supplier Number: 64917-2
- Wafer Container Single84 x 11 mm75 mm (3")Each$163.30Pack of 6$118.47Supplier Number: 64917-3
- Wafer Container Single110 x 11 mm100 mm (4")Each$244.28Pack of 6$186.52Supplier Number: 64917-4
- Wafer Container Single160 x 11 mm150 mm (6")Each$297.30Pack of 6$226.75Supplier Number: 64917-6
- Wafer Container Single170 x 170 x 20 mm100 – 150 mm (4 – 6")Each$385.54Pack of 6$293.49Supplier Number: 64917-46
- Wafer Container Multi—50 mm (2")Each$187.73Pack of 3$141.82Supplier Number: 64918-2
- Wafer Container Multi—75 mm (3")Each$225.56Pack of 3$172.49Supplier Number: 64918-3
- Wafer Container Multi—100 mm (4")Each$398.07Pack of 3$301.86Supplier Number: 64918-4