Disposable Scalpel Sets, Electron Microscopy Sciences
Disposable Scalpel Sets, Electron Microscopy Sciences
Disposable Scalpel Sets, Electron Microscopy Sciences
Certificates
Disposable Scalpel Sets, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
High quality carbon steel is mounted on plastic handle, individually foil wrapped and sterilized by gamma irradiation.
They are exceptionally strong and relatively uniformity sharp, ideal for routine laboratory dissecting, tissue mincing and where you do not need high cost blades.
Packaging: Packed 10 each /box.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • Scalpel Handle Set #10
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $29.85
    $29.85
    Log in for availability
    Pack of 10
    Pack of 10 - $19.80
    $19.80
    Each
       $29.85
    Pack of 10
       $19.80
    Supplier Number: 72052-10
  • Scalpel Handle Set #11
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $29.85
    $29.85
    Log in for availability
    Pack of 10
    Pack of 10 - $19.80
    $19.80
    Each
       $29.85
    Pack of 10
       $19.80
    Supplier Number: 72052-11
  • Scalpel Handle Set #15
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $29.85
    $29.85
    Log in for availability
    Pack of 10
    Pack of 10 - $19.80
    $19.80
    Each
       $29.85
    Pack of 10
       $19.80
    Supplier Number: 72052-15
  • Scalpel Handle Set #20
    Discontinued
    Supplier Number: 72052-20
  • Scalpel Handle Set #22
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $29.48
    $29.48
    Log in for availability
    Pack of 10
    Pack of 10 - $19.54
    $19.54
    Each
       $29.48
    Pack of 10
       $19.54
    Supplier Number: 72052-22