Cryostat Frozen Sectioning Aid System
À propos de cet article
Frozen sections created as thin as two microns with the same quality as sections cut from paraffin blocks.
- Preserves sample morphology
- Increases quality and reduces repeats
- Fast and effortless serial sectioning
- Sections can be fixed anhydrously
The CryoJane® tape-transfer system creates frozen sections as thin as 2 µm with the same quality as sections cut from paraffin blocks. Sections are wrinkle-free, uncompressed, and fully intact when bonded to the microscope slide. The Cryogen uses adhesive coated slides and adhesive tapes to capture sections instead of using an anti-roll plate or brush.
With this system, thin tissue sections remain unflawed even after mounting. This makes Cryogen ideal for applications that require perfectly thin sections to clarify structural relationships and reveal morphological detail.
The CryoJane tape-transfer system is adaptable to most cryostats on the market. Sections can be fixed 'anhydrously' to preserve virtually all fine structures present in the snap-frozen block of tissue. Water-soluble enzymes, antigens and nucleo-proteins are preserved in situ. With appropriate fixation and staining, true localization of enzyme and antigen activity can be visualized. The bond between the section and the slide is solvent resistant and permanent, preventing section loss during staining.
CryoJane is suitable for routine cryosections and is indispensable for sectioning difficult tissues including fatty breast and undecalcified bone. Preserves sample morphology. Increases quality, reduces repeats and streamlines workflows. Allows fast and virtually effortless serial sectioning.
: CE and UL Listed.
: The Leica CryoJane tape-transfer system includes the control unit, (ECU) with a power cord, UV flash unit with temperature controlled pad, hand roller and mounting, bracket, door bar, five cable clamps, one slide storage shelf, 125 1X adhesive slides, 10¹/₂X adhesive slides, 10 4X adhesive slides, one roll of tape windows (400 tapes per roll), a manual, one set of replacement fuses, knife facet wipers and a one year parts warranty from date of install.
2 Options Available Below
Product Details & Documents
Frozen sections created as thin as two microns with the same quality as sections cut from paraffin blocks.
- Preserves sample morphology
- Increases quality and reduces repeats
- Fast and effortless serial sectioning
- Sections can be fixed anhydrously
The CryoJane® tape-transfer system creates frozen sections as thin as 2 µm with the same quality as sections cut from paraffin blocks. Sections are wrinkle-free, uncompressed, and fully intact when bonded to the microscope slide. The Cryogen uses adhesive coated slides and adhesive tapes to capture sections instead of using an anti-roll plate or brush.
With this system, thin tissue sections remain unflawed even after mounting. This makes Cryogen ideal for applications that require perfectly thin sections to clarify structural relationships and reveal morphological detail.
The CryoJane tape-transfer system is adaptable to most cryostats on the market. Sections can be fixed 'anhydrously' to preserve virtually all fine structures present in the snap-frozen block of tissue. Water-soluble enzymes, antigens and nucleo-proteins are preserved in situ. With appropriate fixation and staining, true localization of enzyme and antigen activity can be visualized. The bond between the section and the slide is solvent resistant and permanent, preventing section loss during staining.
CryoJane is suitable for routine cryosections and is indispensable for sectioning difficult tissues including fatty breast and undecalcified bone. Preserves sample morphology. Increases quality, reduces repeats and streamlines workflows. Allows fast and virtually effortless serial sectioning.
: CE and UL Listed.
: The Leica CryoJane tape-transfer system includes the control unit, (ECU) with a power cord, UV flash unit with temperature controlled pad, hand roller and mounting, bracket, door bar, five cable clamps, one slide storage shelf, 125 1X adhesive slides, 10¹/₂X adhesive slides, 10 4X adhesive slides, one roll of tape windows (400 tapes per roll), a manual, one set of replacement fuses, knife facet wipers and a one year parts warranty from date of install.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Adhesive slides, 1x coating 1Discontinued-Supplier Number: -
- Adhesive slides, 4x coatingDiscontinued-Supplier Number: -
- Fuses - 1-AmpDiscontinued-Supplier Number: -
- Aqueous buffer salt mixDiscontinued-Supplier Number: -
- Knife facet wipersDiscontinued-Supplier Number: -
- Adhesive slides,¹/₂x coating 1Each$995.75Pack of 125$837.02Supplier Number: 62800-X
- Block holderDiscontinued-Supplier Number: -
- Supplier Number: 62800-44
- Fuses - 1-AmpDiscontinued-Supplier Number: -
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Tape Windows
Informations sur le produit
- Catalog #AvailabilityPrice
- Regular Adhesive Slide, 1x
Informations sur le produit
- Catalog #AvailabilityPrice
- Hand Roller