Extra Deep Trays, Electron Microscopy Sciences
À propos de cet article
These deep trays with dividers, can organize your larger and bulkier supplies.
- Measures L×W: 17.5×19.5"
- With heights of either 4, 6, or 8"
They are constructed of bright white plastic with smooth surface.
Available with three depths: 4, 6, and 8 inches, and with or without hinged clear lid.
1 Options Available Below
Product Details & Documents
These deep trays with dividers, can organize your larger and bulkier supplies.
- Measures L×W: 17.5×19.5"
- With heights of either 4, 6, or 8"
They are constructed of bright white plastic with smooth surface.
Available with three depths: 4, 6, and 8 inches, and with or without hinged clear lid.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 64367-4
- Discontinued-Supplier Number: -