Bacterial Cell Spreader, Electron Microscopy Sciences
Product Details & Documents
Made from high impact polystyrene. No flame sterilization needed. Designed for easy spreading of cells onto the surface of an agar plate. Supplied sterile in individually packs. Total length of handle: 176 mm, width of spreader: 36 mm. Color green.
Specifications
- Désignation:Bacterial Cell Spreader