Antibody Saver Tray, Electron Microscopy Sciences
Fournisseur: Electron Microscopy Sciences
Certificats
À propos de cet article
Antibody saver trays minimize the use of expensive antibodies when incubating filters in the Western Blot process.
- Trays are available for popular midi and mini and multiple lane filters
- Each tray is molded of polypropylene and includes an attached lid to protect from spillage
- Trays are stackable, reclosable and reusable
- Single lane trays are linked together for easier handling but can be separated
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together