Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Parafilm Dispenser, Electron Microscopy Sciences
Store, dispense, and cut Parafilm®, tape, labels, and Tough-Tags. Unique design for maximum safety. ABS plastic dispenser designed with a safety razor that slides and cuts with no danger to your fingers.
  • Holds one or two 2" rolls, or one 4" roll
  • Size (W x H x D): 120 x 156 x 171 mm
  • Parafilm is a registered trademark of American can company
Product Family Options
  • Désignation
  • Parafilm Dispenser Blue
    Each
    Each - $155.58
    $155.58
    Each
       $155.58
     : 70993-01
  • Parafilm Dispenser Green
    Each
    Each - $160.82
    $160.82
    Each
       $160.82
     : 70993-02
  • Parafilm Dispenser Red
    Each
    Each - $183.91
    $183.91
    Each
       $183.91
     : 70993-03
  • Replacement Blades for Dispenser
    Each
    Each - $27.14
    $27.14
    Pack of 5
    Pack of 5 - $20.67
    $20.67
    Each
       $27.14
    Pack of 5
       $20.67
     : 70993-04