Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
Certificates
Phosphate Buffered Saline, Electron Microscopy Sciences
Phosphate Buffered Saline, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
PBS is commonly used in biochemistry.
It is a salty solution containing calcium chloride, sodium phosphate and potassium phosphate. PBS is isotonic and non-toxic to cells. 1X PBS final concentration is 8.0 g/l NaCl, 0.2 g/l KCl, 1.42 g/l Na₂HPO₄, 0.24 g/l KH₂PO₄ and pH 7.4.
Product Family Options
Product Information
  • Désignation
    Taille
    Type de conditionnement
    Availability
    Price
  • Phosphate Buffered Saline, 10X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19342-10
  • Phosphate Buffered Saline, 2X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19343-10
  • Phosphate Buffered Saline, 1X
    1 l
    Bottle
    Discontinued
    Supplier Number: 19344-10
  • Phosphate Buffered Saline, 10X
    3.78 l
    Bottle
    Log in for availability
    Log in for availability
    Each
    Each - $333.24
    $333.24
    Each
       $333.24
    Supplier Number: 19242-40
  • Phosphate Buffered Saline, 2X
    3.78 l
    Bottle
    Discontinued
    Supplier Number: 19343-40
  • Phosphate Buffered Saline, 1X
    3.78 l
    Bottle
    Discontinued
    Supplier Number: 19344-20