SEM Single Mount Storage Tube and Mailer, Hitachi, Electron Microscopy Sciences
Ideal for storage as well as mailing.
A 15 mm diamter plastic tube and plug is for M4 threaded Hitachi mounts. A 15 mm Hitachi mount can be inserted securely to the plug and then protected by the tube. The internal diameter of the tube is 21 mm (0.83”). The Outer measurements of the tube are: 23 x 52 mm (0.9 x 2.05").
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together