Rectangular Stain Dish, Electron Microscopy Sciences
Rectangular Stain Dish, Electron Microscopy Sciences
Rectangular Staining Dish, 3 x 2 1/4 x 1 1/4
Catalog # 102096-606
Supplier #:  70314-01
Rectangular Stain Dish, Electron Microscopy Sciences
Catalog # 102096-606
Rectangular Stain Dish, Electron Microscopy Sciences
Rectangular Stain Dish, Electron Microscopy Sciences
Rectangular Staining Dish, 3 x 2 1/4 x 1 1/4
Price
$25.79
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Heavy, clear, soda lime glass stain dish, accept glass slide measure 75 x 25 mm or 3 x 1”.
This dish comes with glass cover, holds 20 slides back-to-back horizontally.
Inside dimensions: 3 x 2 1/4x 1 1/4”.
Spécifications
  • Désignation:
    Rectangular Staining Dish
  • Dimensions:
    3 x 2 1/4 x 1 1/4