Indexed Grids, Electron Microscopy Sciences
À propos de cet article
Useful for particle counting and in area of specimen determinations.
- NE10A: numbered grid 5×5 mm, 0.5 mm pitch
- NE11A: numbered grid 10×10 mm, 1.0 mm pitch
- NE34A: numbered grid 1×1 mm, 0.1 mm pitch
- NE71: index pattern 20×20 grid of 0.5 mm squares
8 Options Available Below
Product Details & Documents
Useful for particle counting and in area of specimen determinations.
- NE10A: numbered grid 5×5 mm, 0.5 mm pitch
- NE11A: numbered grid 10×10 mm, 1.0 mm pitch
- NE34A: numbered grid 1×1 mm, 0.1 mm pitch
- NE71: index pattern 20×20 grid of 0.5 mm squares
Product Family Options
Informations sur le produit
- DésignationTailleAvailabilityPrice
- Supplier Number: 68016-16
- Graticle NE11A19 mmDiscontinued-Supplier Number: 68016-19
- Supplier Number: 68016-20.4
- Graticle NE7121 mmDiscontinued-Supplier Number: 67981-21
- Graticle NE34A21 mmDiscontinued-Supplier Number: 68018-21A
- Graticle NE11A21 mmDiscontinued-Supplier Number: 68016-21
- Graticle NE7123 mmDiscontinued-Supplier Number: 67981-23
- Supplier Number: 68016-23Spécifications:MarketsourceMarketsource
- Graticle NE7124 mmDiscontinued-Supplier Number: 67981-24
- Supplier Number: 68016-24.5
- Supplier Number: 68016-25
- Graticle NE7126 mmDiscontinued-Supplier Number: 67981-26
- Supplier Number: 68016-26
- Supplier Number: 68016-27
- Supplier Number: 67981-50
- Graticle NE10A21 mmDiscontinued-Supplier Number: 68011-21A