About this item
This device is used as an aid when programming a microwave oven to achieve an ideal target temperature of 20 – 100 µL staining volumes.
The tool is designed to give quantitative temperature information about the droplet during staining of tissue sections on electron microscope grids. This unit is a 4-well PTFE coated glass slide, each well 8mm in diameter which will maintain the uniform shape of the droplet; 4 liquid crystal temperature strips ranging from 35 to 50° are affixed to each well to monitor the microwave oven temperature.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.