Fluor Adhesive Chamber, Electron Microscopy Sciences
Each package includes illustrated instructions for use, applicator, and 200 adhesive port seals
These new microarray chambers are designed specially for compatibility with Cy5® or Alexa Fluor 647® direct labeled DNA probes for hybridization assays. These chambers as well as the original HybriWells may also be used with all other fluorochrome labeled probes.
: Additional seal tabs are sold separately: Adesive Port Seal Tabs.
Each package includes illustrated instructions for use, applicator, and 200 adhesive port seals
These new microarray chambers are designed specially for compatibility with Cy5® or Alexa Fluor 647® direct labeled DNA probes for hybridization assays. These chambers as well as the original HybriWells may also be used with all other fluorochrome labeled probes.
: Additional seal tabs are sold separately: Adesive Port Seal Tabs.
Product Family Options
- TailleNb de puitsVolumeProfondeur
- 22 mm x 22 mm130 - 50 µl0.25 mm-: 70328-50
- 22 mm x 22 mm115 - 25 µl0.12 mm-: 70328-51
- 22 mm x 40 mm1180 - 200 µl0.25 mm-: 70328-52
- 22 mm x 40 mm190 - 100 µl0.12 mm-: 70328-53
- 22 mm x 60 mm1280 - 330 µl0.25 mm-: 70328-54
- 22 mm x 60 mm1140 - 165 µl0.12 mmEach$1,007.99Pack of 100$768.75: 70328-55
- 21 mm x 40 mm150 - 100 µl0.12 mm-: 70328-56