Solvent Resistant Pen, Electron Microscopy Sciences | Avantor
Solvent Resistant Pen, Electron Microscopy Sciences
Solvent Resistant Pen, Electron Microscopy Sciences
Certificates
Solvent Resistant Pen, Electron Microscopy Sciences
Solvent Resistant Pen, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Solvent Resistant Pens feature an ultra-permanent ink which is resistant to xylene, acetone, and formalin.
Markings will not run on embedding cassettes or frosted/painted slides and labware, even afterrepeated washes and solution treatments. The markings can be removed with acetic acid, an ethanol/xylene mixture or paraffin. Testing advised before use.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Point Size
    Availability
    Price
  • Solvent Resistant Pen
    Black
    Extra Fine (0.6 mm)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.63
    $18.63
    Each
       $18.63
    Supplier Number: 62053-B
  • Solvent Resistant Pen
    Red
    Extra Fine (0.6 mm)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $18.63
    $18.63
    Each
       $18.63
    Supplier Number: 62053-R