About this item
Teeny Writers are perfect general use lab markers
The Twin-Tip Teeny Writer has a fine tip and an ultra fine tip. The original Teeny Writer has an extra fine tip. Both are ideal for writing on Tough-Tags and Tough-Spots. Twin-Tip Too laboratory marking pen provides a new option for all writing needs. It is resistant to most mild laboratory chemicals and gives the flexibility of a twin tip pen for fine tip writing.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.