Flexwell™ Incubation Chambers, Electron Microscopy Sciences
À propos de cet article
FlexWell™ removable incubation chambers for microarrays using ONCYTE® slides.
- Silicone FlexWell™ forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide
- Slides and FlexWell™ fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate
- Gasket may be sealed for mixing and to prevent evaporation
- Available in eight-well and 16-well formats
2 Options Available Below
Product Details & Documents
FlexWell™ removable incubation chambers for microarrays using ONCYTE® slides.
- Silicone FlexWell™ forms well on slides using a clean release adhesive to isolate up to 16 specimens per slide
- Slides and FlexWell™ fit into a reusable slide tray organizing wells into the footprint of a SBS compliant microtiter plate
- Gasket may be sealed for mixing and to prevent evaporation
- Available in eight-well and 16-well formats
Product Family Options
Informations sur le produit
- DésignationØ ext.ProfondeurAvailabilityPrice
- Flexwell 8- 6.5 x 6.5 mm wells, Clear Silicone Adhesive one Side25 x 75 mm3.2 mmDiscontinued-Supplier Number: 63485-50
- Flexwell 16- 6.5 x 6.5 mm wells, Clear silicone25 x 75 mm3.2 mmEach$495.84Pack of 10$378.50Supplier Number: 63485-51
- Flexwell 64- 3.5 x 3.5 mm wells, Clear Silicone25 x 75 mm1.8 mmDiscontinued-Supplier Number: 63485-52
- Seal Strips, 24 x 77 mm Seal Area——Discontinued-Supplier Number: 63485-53
- Supplier Number: 63485-54
- Flexplate, 96 Square Wells, Clear110 x 75 mm4 mmDiscontinued-Supplier Number: 63485-55
- Flexplate, 96 Square Wells, Black110 x 75 mm4 mmDiscontinued-Supplier Number: -