Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames, Electron Microscopy Sciences
Gel Drying Frames allow researchers to quickly and easily dry gels between two sheets of cellophane without the use of expensive heat/vacuum gel dryers.
  • Two open frames
  • Two solid back plates
  • 12 side clips
  • 50 sheets of pre-cut cellophane
The 24 x 24 cm frames allow for drying of larger format gels, while the 14 x 14 cm frames are ideal for mini gels and other smaller formats.
Product Family Options
  • Désignation
  • Gel Drying Frames 14 x 14 cm Gel Frame
    Each
    Each - $347.63
    $347.63
    Kit of 2
    Kit of 2 - $347.63
    $347.63
    Each
       $347.63
    Kit of 2
       $347.63
     : 68510-14
  • Gel Drying Frames 24 x 24 cm Gel Frame
    Each
    Each - $408.43
    $408.43
    Kit of 2
    Kit of 2 - $408.43
    $408.43
    Each
       $408.43
    Kit of 2
       $408.43
     : 68510-24
  • Gel Drying Frames 14 x 14 cm Cellophane
    Each
    Each - $44.36
    $44.36
    Pack of 50
    Pack of 50 - $56.03
    $56.03
    Each
       $44.36
    Pack of 50
       $56.03
     : 68511-14
  • Gel Drying Frames 24 x 24 cm Cellophane
    Each
    Each - $55.67
    $55.67
    Pack of 50
    Pack of 50 - $49.01
    $49.01
    Each
       $55.67
    Pack of 50
       $49.01
     : 68511-24