Specialty Labels, Electron Microscopy Sciences
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Preprinted Tough-Spots make labeling and identifying samples quick and easy.
- Temperature resistant from –196 to 80 °C
Use the numbered Tough-Spots to identify a series of samples - no handwriting or printing needed. Each color coded ToughSpot has a distinctly different color to identify similar samples and also includes a white area to clearly write a quick note about it.
Lab safety is always a top priority. Use the Biohazard or Radioactive Tough-Spots to identify tubes or other vessels containing hazardous materials.
Product Family Options
- DésignationLegendColorDiameter
- Tough-SpotsBiohazard—3/8"Each$94.67Roll of 1,000$94.67: 77593-14
- Tough-SpotsRadioactive—3/8"-: -
- Tough-SpotsBullseyeDark green7/16"-: -
- Tough-SpotsBullseyeCopper7/16"-: 77593-12
- Tough-SpotsBullseyeDark purple7/16"-: -