AMS Sorbothane Balance Platform, Electron Microscopy Sciences
AMS Sorbothane Balance Platform, Electron Microscopy Sciences
Ams sorbothane balance platform
Catalog # 102090-930
Supplier #:  6870
AMS Sorbothane Balance Platform, Electron Microscopy Sciences
Catalog # 102090-930
AMS Sorbothane Balance Platform, Electron Microscopy Sciences
AMS Sorbothane Balance Platform, Electron Microscopy Sciences
Ams sorbothane balance platform
Price
$1,904.57
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The AMS tables incorporate Sorbothane isolators, giving excellent isolation with a natural frequency of less than 7 Hz.
Our isolators considerably outperform both simple air cushion and rubber isolators, shortening stabilization times and speeding up measurements.
Spécifications
  • Désignation:
    Ams sorbothane balance platform