Cell Stretcher, Electron Microscopy Sciences
Product Details & Documents
Cell Stretcher CS-10 Series is a device for simultaneous live cell imaging with motion compensation for uni-axial mechanical straining or compression.
- Cells can be imaged even during stretch
- Stretching chamber allows use of commercially available PDMS sheets
- Cyclic protocols with different characteristics (linear, sinusoidal, sigmoidal) can be programmed
- Position of the membrane chamber along the stretch axis can be changed
This compact cell strain device makes it possible to perform life cell imaging studies where a rapid image acquisition is required, such as observing changes of the intracellular Ca2+ concentration when cells are mechanically stimulated.
Specifications
- Désignation:Membrane for Cell Stretcher