Hitachi SEM Mount Box, Electron Microscopy Sciences
EMS-Hitachi SEM Mount Box
  102091-312
Hitachi SEM Mount Box, Electron Microscopy Sciences
  102091-312
Hitachi SEM Mount Box, Electron Microscopy Sciences
Hitachi SEM Mount Box, Electron Microscopy Sciences
EMS-Hitachi SEM Mount Box
$32.05
Each
EMS introduces a unique box  for Hitachi SEM mounts with M4 threaded hole in the base
The box is numbered 1 – 10 to accommodate Hitachi mounts from 15 to 32 mm diameter.
  • Désignation:
    EMS-Hitachi SEM Mount Box