ProPlate™ MicroArray System, Electron Microscopy Sciences
À propos de cet article
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
1 Options Available Below
Product Details & Documents
ProPlate™ MicroArray System is an integration of microplate and biochip technologies
Each ProPlate Tray Set includes: 4 Slide modules, 8 Snap clips, 1 Tray and Cover, 10 seal strips and one applicator. Tray conforms to SBS standards for footprint (81.7mm width x 123.7mm length) and well spacing. System available in Chip Chip™ compatible formats.
ProPlate™ integrates microscope slide based array technology with automated microtiter plate processing. Up to 64 individual arrays can be assayed per slide. Four slides fit into a tray, producing a modular plate with a standard microtiter plate footprint and well spacing allowing automated robotic processing up to 256 arrays.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- 1 Well ProPlate Tray Set, 3-16.5x70 mm Well, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-01
- 2 Well ProPlate Slide Module Tray Set, 3-16.5x34 mm Well, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-02
- 3 Well ProPlate Slide Module Tray Set, 3-18x20 mm Well, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-03
- 4 Well ProPlate Slide Module Tray Set, 3-16x16 mm Well, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-04
- 8 Well ProPlate Slide Module Tray Set, 3-16x16 mm, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-05
- 16 Well ProPlate Slide Module Tray Set, 3-6 mm dia, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-06
- 16 Well ProPlate Tray Set 3-7x7 mm, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-07
- 64 Well ProPlate Slide Module Tray Set 3-3.5x3.5 mm, Stainless Steel Spring ClipsDiscontinued-Supplier Number: 63484-08
- 1 Well ProPlate Module Tray Set 4-16.5x70 mm Well, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-09
- 2 Well ProPlate Module Tray Set, 4-16.5x34 mm Well, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-10
- 3 Well ProPlate Module Tray Set 4-18x20 mm, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-11
- 4 Well ProPlate Module Tray Set, 4-16x16 mm, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-12
- 8 Well ProPlate Slide Module Tray Set, 4-7x16 mm, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-13
- 16 Well ProPlate Slide Module Tray Set, 4-6 mm dia, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-14
- 16 Well ProPlate Slide Module Tray Set, 4-7x7 mm, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-15
- 64 Well ProPlate Slide Module Tray Set, 4-3.5x3.5 mm, Delrin Snap ClipsDiscontinued-Supplier Number: 63484-16
- 1 Well Module, 16.5x70 mm WellDiscontinued-Supplier Number: 63484-21
- 2 Well Module, 16.5x34 mm WellDiscontinued-Supplier Number: 63484-22
- 3 Well Module, 18x20 mm WellDiscontinued-Supplier Number: 63484-23
- 4 Well Module, 16x16 mm WellDiscontinued-Supplier Number: 63484-24
- 8 Well Module, 7x16 mm WellDiscontinued-Supplier Number: 63484-25
- 16 Well Module, 6 mm dia WellDiscontinued-Supplier Number: 63484-26
- 16 Well Module, 6x7 mm WellDiscontinued-Supplier Number: 63484-27
- 64 Well Module, 3.5x3.5 mm WellDiscontinued-Supplier Number: 63484-28
- 1 Well Module, 16.5x70 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-31
- 2 Well Module, 16.5x34 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-32
- 3 Well Module, 18x20 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-33
- 4 Well Module 16x16 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-34
- 8 Well Module, 7x16 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-35
- 16 Well Module, 6 mm dia Well, Delrin ClipDiscontinued-Supplier Number: 63484-36
- 16 Well Slide Module, 7 x 7 mm, Delrinl ClipsDiscontinued-Supplier Number: 63484-37
- 64 Well Module, 3.5x3.5 mm Well, Delrin ClipDiscontinued-Supplier Number: 63484-38
- 1 Well Module, 16.5x70 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-41
- 2 Well Module, 16.5x34 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-42
- 3 Well Module, 18x20 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-43
- 4 Well Module, 16x16 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-44
- 8 Well Module, 7x16 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-45
- 16 Well Module, 6 mm dia Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-46
- 16 Well Module, 6x7 mm Well,Ss ClipDiscontinued-Supplier Number: 63484-47
- 64 Well Module, 3.5x3.5 mm Well, Stainless Steel ClipDiscontinued-Supplier Number: 63484-48
- 1 Well Silicone Gasket, 16.5x70 mmDiscontinued-Supplier Number: 63484-51
- 2 Well Silicone Gasket, 16x5x24 mmDiscontinued-Supplier Number: 63484-52
- 3 Well Silicone Gasket, 16x20 mmDiscontinued-Supplier Number: 63484-53
- 4 Well Silicone Gasket, 16x16 mmDiscontinued-Supplier Number: 63484-54
- 8 Well Silicone Gasket, 7x16 mmEach$288.78Pack of 10$212.49Supplier Number: 63484-55
- 16 Well Silicone Gasket, 6 mm diaDiscontinued-Supplier Number: 63484-56
- 16 Well Propalte Silicone Gasket, 7x7 mmDiscontinued-Supplier Number: 63484-57
- 64 Well Silicone Gasket, 3.5x3.5 mmDiscontinued-Supplier Number: 63484-58
- Seal Strips & Seal Strip ApplicatorDiscontinued-Supplier Number: 63484-80
- Delrin Non Numbered Snap ClipsDiscontinued-Supplier Number: 63484-81
- Stainless Steel Snap ClipsDiscontinued-Supplier Number: 63484-82
- Tray & Cover for Snap Clip ModulesDiscontinued-Supplier Number: 63484-83
- Tray & Cover for Spring Clip ModulesDiscontinued-Supplier Number: 63484-84