Labeling Tape, Electron Microscopy Sciences
À propos de cet article
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
10 Options Available Below
Product Details & Documents
Use pen, pencil or marker to mark. Leaves no adhesive residue when peeled off.
Water, oil, and acid resistant. Withstands cold (−23 °C) and hot (121 °C) temperatures.
Product Family Options
Informations sur le produit
- ColorTailleAvailabilityPrice
- Blue0.75" x 500"Each$38.31Roll of 1$38.31Supplier Number: 77627-B
- Green0.75" x 500"Each$38.31Roll of 1$38.31Supplier Number: 77627-G
- Red0.75" x 500"Each$38.31Roll of 1$38.31Supplier Number: 77627-R
- White0.75" x 500"Each$32.49Roll of 2$32.49Supplier Number: 77627-W
- Yellow0.75" x 500"Each$37.41Roll of 1$37.41Supplier Number: 77627-Y
- Blue0.75" x 2160"Each$93.14Roll of 1$93.14Supplier Number: 77628-B
- Green0.75" x 2160"Each$109.43Roll of 1$109.43Supplier Number: 77628-G
- Red0.75" x 2160"Each$109.43Roll of 1$109.43Supplier Number: 77628-R
- White0.75" x 2160"Each$109.43Roll of 1$109.43Supplier Number: 77628-W
- Yellow0.75" x 2160"Each$109.43Roll of 1$109.43Supplier Number: 77628-Y