Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, Electron Microscopy Sciences
Certificates
Write Protect Tape, Electron Microscopy Sciences
Write Protect Tape, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Write Protect is a clear adhesive tape designed to cover and protect writing or printing on labels, on the surface of microtubes and on other laboratory containers.
  • Will not dry out, peel or fall off
  • Withstand temperatures ranging from –196 to 150 °C
The clear tape comes in a handy dispenser and will adhere to all plastics, glass, metals and label materials. The tape is provided in perforated, pull-off segments for 0.5 and 1.5 mL tubes.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Availability
    Price
  • Write Protect Tape
    32 x 13 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.34
    $87.34
    Log in pour la disponibilité
    Roll of 1,000
    Roll of 1,000 - $87.34
    $87.34
    Each
       $87.34
    Roll of 1,000
       $87.34
    Supplier Number: 77626-10
  • Write Protect Tape
    41 x 16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $87.34
    $87.34
    Log in pour la disponibilité
    Roll of 1,000
    Roll of 1,000 - $87.34
    $87.34
    Each
       $87.34
    Roll of 1,000
       $87.34
    Supplier Number: 77626-20