Autoradiography Ruler, Electron Microscopy Sciences
Autoradiography ruler
  102096-324
 :  68550-04
Autoradiography Ruler, Electron Microscopy Sciences
  102096-324
Autoradiography Ruler, Electron Microscopy Sciences
Autoradiography Ruler, Electron Microscopy Sciences
Autoradiography ruler
$102.87
Each Pack of 4
The Autoradiography Ruler Set (ARS) is an orientation and measurement device that exposes metric ruler markings directly onto X-ray film.
  • Phosphorescent, non-radioactive metric rulers for radiographs
  • Make easy electrophoretic mobility calculations
Utilizes a specially formulated phosphorescent dye compound and a unique emission filter which serves as an ideal non-radioactive substitute for ink markings. The phosphorescent emission of each ruler can be regenerated indefinitely and can be reused any number of times. Produces a high resolution image that is unaffected by intensifying screens or pre-flashed film.
  • Désignation:
    Autoradiography ruler