Ice Pan, Electron Microscopy Sciences
Ice Pan, Electron Microscopy Sciences
Ice Pan, Electron Microscopy Sciences
Ice Pan, Electron Microscopy Sciences
Ice Pan, Electron Microscopy Sciences
*Not recommended for long-term exposure to harsh chemicals or corrosives.
Half the price of competitor's ice pans. Thick pan walls will not sweat or leave puddles of condensation on the benchtop. Made of triple density expanded polystyrene creating a durable container that is virtually unbreakablePreserves ice up to 30% longer than conventional ice pans. Designed with an inner edge enabling the pans to stack on top of one another for easy storage and to minimize bench-top space. Ice pans are extremely lightweight (1.1, 4.7. and 11.4 oz.) and easy to handle. They feature a molded lip around the edge for carefree moving and carrying. Resistant* to mild chemicals as well as ice-alcohol slurry, dry ice and liquid nitrogen.
*Not recommended for long-term exposure to harsh chemicals or corrosives.
Product Family Options
  • Désignation
    Capacité
    Largeur
    Profondeur
    Hauteur
  • Ice pan, large
    9 L
    17.25"
    13.50"
    4.96"
    Each
    Each - $162.73
    $162.73
    Each
       $162.73
     : 63311-1L
  • Ice pan, medium, triple
    4 L
    12.25"
    8.75"
    4.75"
    Each
    Each - $135.93
    $135.93
    Each
       $135.93
     : 63311-1M
  • Ice pan, small
    1 L
    6.13"
    6.13"
    3.61"
    Each
    Each - $62.34
    $62.34
    Each
       $62.34
     : 63311-1S
  • 13.50"
     : -
  • Ice pan, medium, triple
    4 L
    12.25"
    8.75"
    4.75"
     : -
  • 6.13"
     : -