Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences | Avantor
Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
Certificates
Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
Rota-Rack Rotating Tube Rack, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Each module of the Rota-Rack for large tubes holds two 50 mL tubes, eight 15 mL tubes, six 20 mm tubes, or ten 10-13 mm tubes
Connect up to 4 interlocking racks. Twist and turn to suit your application. Rotate each of the four modules independently. Use several sizes of test tubes, culture tubes, centrifuge tubes, or microcentrifuge tubes at the same time. Click each module into place, load with tubes, and carry. Rack maintains its orientation until you twist it again. Comes fully assembled. Autoclavable.
Each module of the Rota-Rack for large tubes holds two 50 mL tubes, eight 15 mL tubes, six 20 mm tubes, or ten 10-13 mm tubes. Each module of the Rota Rack for microtubes holds six 15 mL tubes, nine 1.5/2.0 mL tubes, twelve 0.5/0.6 mL tubes, or thirty-two 0.2 mL PCR* tubes or four 8-tube strips.
Each module of the Rota Rack for microtubes holds six 15 mL tubes, nine 1.5/2.0 mL tubes, twelve 0.5/0.6 mL tubes, or thirty-two 0.2 mL PCR* tubes or four 8-tube strips.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Rota-Rack for Large Tubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $191.59
    $191.59
    Each
       $191.59
    Supplier Number: 61604-10
  • Rota-Rack for Microtubes
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $196.24
    $196.24
    Each
       $196.24
    Supplier Number: 61604-20