100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
Certificates
100-Well Storage Racks, Electron Microscopy Sciences
100-Well Storage Racks, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Polypropylene rack features durable three point hinges and clasp for secure lid closure.
  • Holds 1.5 to 2.0 ml microtubes and cryovials
  • Rack measures 5.625×5.875×2.25"
User friendly rack features molded gridlines on the lid, imprinted coordinates on the bottom of base and imprinted and raised coordinates on the box.
Product Family Options
Product Information
  • Color
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $153.04
    $153.04
    Log in for availability
    Pack of 5
    Pack of 5 - $108.25
    $108.25
    Each
       $153.04
    Pack of 5
       $108.25
    Supplier Number: 61601-B
  • Natural
    Log in for availability
    Log in for availability
    Log in for availability
    Each
    Each - $144.71
    $144.71
    Log in for availability
    Pack of 5
    Pack of 5 - $102.35
    $102.35
    Each
       $144.71
    Pack of 5
       $102.35
    Supplier Number: 61601-N