81-Well Storage Rack, Electron Microscopy Sciences
À propos de cet article
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
- Molded grid lines on the lid
- Imprinted coordinates on the bottom of base
- Molded reference point on the lid
- Rack measures 5.125×5.125×1.75 cm
5 Options Available Below
Product Details & Documents
Durable polypropylene rack fits in standard 5.25” square freezer modules and holds 1.5 to 2.0 ml microtubes and cryovials.
- Molded grid lines on the lid
- Imprinted coordinates on the bottom of base
- Molded reference point on the lid
- Rack measures 5.125×5.125×1.75 cm
Product Family Options
Informations sur le produit
- DésignationColorAvailabilityPrice
- 81-Well Storage RackAssorted colorsEach159,15 $Pack of 5134,19 $Supplier Number: 61600-A
- 81-Well Storage RackBlueDiscontinued-Supplier Number: 61600-B
- 81-Well Storage RackGreenEach157,66 $Pack of 5132,91 $Supplier Number: 61600-G
- 81-Well Storage RackNaturalEach158,36 $Pack of 5133,53 $Supplier Number: 61600-N
- 81-Well Storage RackOrangeDiscontinued-Supplier Number: 61600-O
- 81-Well Storage RackPinkEach159,92 $Pack of 5134,82 $Supplier Number: 61600-P
- 81-Well Storage RackYellowEach157,66 $Pack of 5132,91 $Supplier Number: 61600-Y