Media Disruptor Genie, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Rapid hands-free disruption of up to 12 microtubes (1.5 ml or 2.0 ml) at one time.
- Unique agitation, vortexing, and collision actions for cell disruption and sample resuspension
- Maintain operating speed for up to 15 minutes or continuous operation, even in cold rooms
- Supplied pop-up cup easily attaches for use as a standard single-tube vortex mixer
- Ideal for glass bead procedures using Disruptor Beads
- Less costly than ultrasonic units or homogenizes
- Spherecal lead-free soda lime glass disruptor beads are available in two sizes
- Beads can be used for disruption of yeast, bacteria, and other samples
- Beads can be used in combination with Disruptor Genie or TurboMix™ attachments for Vortex-Genie™ 2, Vortec-Genie™ 2T or Digital Vortex-Genie™ 2
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together