LockMailer Microscope Slide Jar, Electron Microscopy Sciences | Avantor
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
Certificates
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
LockMailer Microscope Slide Jar, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Tamperproof multi purpose container for mailing, staining or storing microscope slides. Constructed of extra strong and opaque clear polypropylene, and high density polyethylene leakproof tamper evident screw cap (The LockMailer can also be used without the tamper evident feature by not used the lock tab), it will hold up to 4 standard 3 x 1” or 25 x 75mm slides vertically.
Inside chambers are slotted to keep slides safely separated. For color coding purposes, use Capinsert™ (#61805-series) that may be inserted on top of closure. This LockMailer™ can be used as small staining vessel. This unit is designed for maximum stability on a bench top with liquid volume only 14 ml. Each standard volume of one slide is 2 ml, therefore when you want to stain 4 slides in this vessel you need only 6 ml of staining solution. Color coded CapInsert™ is available for the LockMailer™. Measures 35 x87 mm H (1 3/8” x 3 7/16”H). Packaging: 100 per pack. 
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Slide Jar Lockmailer
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $124.33
    $124.33
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $112.80
    $112.80
    Each
       $124.33
    Pack of 100
       $112.80
    Supplier Number: 71406-10
  • Capinsert Lilac
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.53
    $15.53
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $10.06
    $10.06
    Each
       $15.53
    Pack of 100
       $10.06
    Supplier Number: 61806-L
  • Capinsert Assorted Colors
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.74
    $15.74
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $10.19
    $10.19
    Each
       $15.74
    Pack of 100
       $10.19
    Supplier Number: 61806-AS
  • Capinsert white
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $15.74
    $15.74
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $10.19
    $10.19
    Each
       $15.74
    Pack of 100
       $10.19
    Supplier Number: 61806-W