ContactUsLinkComponent
U-tek® Ice Packets, Electron Microscopy Sciences
U-tek® Ice Packets, Electron Microscopy Sciences
Catalog # 102093-562
undefined
U-tek® Ice Packets, Electron Microscopy Sciences
Catalog # 102093-562
Supplier Number:  61533-06
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Model 406J, Single U-tek® Ice Packets
  • Cat. no.:
    102093-562
  • Supplier No.:
    61533-06
Specifications
About this item
U-tek® Ice Packets are cushion fragile shipments which won’t dilute ice baths or damage samples
The packets may be refrozen and reuse, withstands pressure of 60 lb per square inch for 5 minutes or longer. The refrigerant contains neutral pH distilled deionized water and proprietary materials certified by the US Food and Drug Administration as food grade chemicals.
-1°C / +30°F
They remain frozen longer than regular ice without the melting mess.