Cassette Opener, Electron Microscopy Sciences
Cassette Opener
  102681-132
 :  65231-01
Cassette Opener, Electron Microscopy Sciences
  102681-132
Cassette Opener, Electron Microscopy Sciences
Cassette Opener, Electron Microscopy Sciences
Cassette Opener
$166.11
Each
Heavy duty stainless steel cassette opener. Works with 25 mm, 37 mm, and Air-O-Cell cassettes. No need to remove labels, this opener cuts right through and effortlessly pries cassettes open.
  • Désignation:
    Cassette Opener