Air Sampling Cassette, 37 mm, Electron Microscopy Sciences
À propos de cet article
The 37 mm Air Sampling Cassette w/MCE Filters are designed to meet all NIOSH, OSHA, and EPA requirements for air sampling standards. All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations.
- Polystyrene housing, 2 or 3 piece
- 3 piece model suitable for lead sampling
- Preloaded, ready-to-use
- Leak-free design
All 37 mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads.
2 Options Available Below
Product Details & Documents
The 37 mm Air Sampling Cassette w/MCE Filters are designed to meet all NIOSH, OSHA, and EPA requirements for air sampling standards. All 37 mm cassette housings are manufactured in house and are available in both two and three piece configurations.
- Polystyrene housing, 2 or 3 piece
- 3 piece model suitable for lead sampling
- Preloaded, ready-to-use
- Leak-free design
All 37 mm cassettes are individually assembled and preloaded under stringent quality assurance standards with only the finest quality membrane and support pads.
Product Family Options
Informations sur le produit
- DésignationTaille de poreAvailabilityPrice
- 37 mm Cassette, MCE Filters, 3 Piece0.8 µmEach$204.95Pack of 50$158.40Supplier Number: 65218-38
- 37 mm Cassette, MCE Filters, 3 Piece0.45 µmEach$245.25Pack of 50$190.44Supplier Number: 65218-45